Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Nitrite 95.0+%, TCI America™
SDP

Catalog No. N037625ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Catalog No. N037625ML

Supplier: TCI America N037625ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 544-16-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002058
InChI Key JQJPBYFTQAANLE-UHFFFAOYSA-N
Synonym 1-butyl nitrite, nitrous acid, butyl ester, n-butyl nitrite, nitrous acid, n-butyl ester, nitrous acid butyl ester, n-butylnitrite, butylnitrite, unii-44p8qg0f3t, ccris 838, n-c4h9ono
PubChem CID 10996
IUPAC Name butyl nitrite
SMILES CCCCON=O

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 mL
UN Number 2351
Formula Weight 103.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Nitrite
EINECSNumber (2)-1565
RTECSNumber RA0780000
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.