Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Butyl Ethyl Ether 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 92°C |
| Quantity | 25 mL |
| UN Number | 1179 |
| Formula Weight | 102.18 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Butyl Ethyl Ether |
Safety and Handling
| RTECSNumber | KN4725000 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction