Learn More
New England Biolabs, Inc. BseRI – 200 units
Supplier: New England Biolabs, Inc. R0581S
A restriction endonuclease that recognizes the sequence GAGGAGNNNNNNNN_NN^.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.