Learn More
Medchemexpress LLC HY-N3014 5mg Medchemexpress, Bruceine D CAS:21499-66-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN30145MG
Encompass_Preferred
Medchemexpress, HY-N3014 5mg Bruceine D CAS:21499-66-1 Bruceine D is a Notch inhibitor with anti-cancer activity and induces apoptosis in several human cancer cells. Bruceine D is an effective botanical insect antifeedant with outstanding systemic properties, causing potent pest growth inhibitory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0409
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.