Learn More
Medchemexpress LLC HY-106048 10mg Medchemexpress, Bredinin aglycone CAS:56973-26-3 Purity:>98%
Supplier: Medchemexpress LLC HY10604810MG
Medchemexpress, HY-106048 10mg Bredinin aglycone CAS:56973-26-3 Bredinin aglycone (5-Hydroxy-1H-imidazole-4-carboxamide) is a purine nucleotide analogue. Bredinin aglycone can be used to examine the efficiency of catalysts for the preparation of purine nucleotide analogues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.