Learn More
BrandTech™ VACUUBRAND™ Rotary Vane Vacuum Pumps: Inlet Separators
Catalog No. p-4673906 Shop All BrandTech Scientific Products
Change view
Click to view available options
For Use With:
RE2, RZ2, RE 2.5, RZ 2.5
RE5, RZ5, RE6, RZ6, RE 2.5, RZ 2.5
RE8, RE9, RE16, RZ8, RZ9, RZ16
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
13-889-109 RE2, RZ2, RE 2.5, RZ 2.5
13-889-112 RE5, RZ5, RE6, RZ6, RE 2.5, RZ 2.5
13-889-113 RE8, RE9, RE16, RZ8, RZ9, RZ16
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-889-109 Supplier BrandTech™ VACUUBRAND™ Supplier No. 20698000
Only null left
Add to Cart
Add to Cart
Inlet separators for VACUUBRAND™ Rotary Vane Vacuum Pumps.
  • For use with VACUUBRAND™ Rotary Vane Vacuum Pumps
  • Inlet Separators 'AK' help keep condensates and particulates from entering the vacuum pump, preserving performance.
Note: For RE/RZ 2.5: For rotary vane pumps RE 2.5 (article no. 2079715x) and RZ 2.5 (article no. 2079812x) - please order Inlet Separator 20698006. For rotary vane pumps RE 2.5 (article no. 2069715x) and RZ 2.5 (article no. 2069812x) - please order Inlet Separator 20698000.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) To keep condensates and particulates from entering the vacuum pump
For Use With RE2, RZ2, RE 2.5, RZ 2.5
Model 20698000
Type Inlet Catchpot/Separator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov