Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SDS Elements Signs

Catalog No. p-4932761 Shop All Brady Corporation Products
Click to view available options
Material:
Fiberglass
Polyester

Catalog No. 19-000-480

Supplier: Brady™ 133172

Only null left
Add to Cart
Add to Cart

To identify equipment, rooms and materials, which contain or are contaminated with hazardous agents

  • Available in fiberglass and self-sticking polyester material
  • Size: 25.4W x 35.5cm H(10 x 14 in.)

Specifications

Material Polyester
Quantity 1 Ea.
Description Self-sticking Polyester (B-302)
Height (English) 14 in.
Height (Metric) 35.5 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Dimensions (L x W) 25.4 x 35.5cm
Displayed Text SDS Format
Product Type SDS Elements Sign
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.