Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron triiodide, 95%

Catalog No. AA8722003
Click to view available options
Quantity:
1 g
5 g

Catalog No. AA8722003

Supplier: Thermo Scientific Chemicals 08722003

Only null left
Add to Cart

CAS: 13517-10-7 | BI3 | 391.52 g/mol

Boron triiodide is used as pharmaceutical, chemical and organic intermediate. It is also used as a catalyst and for preparation of boron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13517-10-7
Molecular Formula BI3
Molecular Weight (g/mol) 391.52
MDL Number MFCD00036286
InChI Key YMEKEHSRPZAOGO-UHFFFAOYSA-N
Synonym boron triiodide, borane, triiodo, triiodoboron, acmc-1bo2g, boron triiodide 1g, inchi=1/bi3/c2-1 3
PubChem CID 83546
IUPAC Name triiodoborane
SMILES IB(I)I

Specifications

Boiling Point 210°C
Melting Point 48°C to 51°C
Color White
Physical Form Crystals
Quantity 1 g
Assay Percent Range >98%
Solubility Information Reacts with water.
Formula Weight 391.52
Odor Pungent
Sensitivity Light and moisture sensitive
Density 3.35
Chemical Name or Material Boron triiodide
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 236-857-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.