Learn More
Bachem Boc-Glu(OBzl)-Gly-Arg-AMC · HCl
Supplier: Bachem 4016428.0050
Boc-Glu(OBzl)-Gly-Arg-AMC · HCl, 133448-22-3, C35H45N7O9 · HCl, Mr 744.25, 50mg. Boc-EGR-AMC, substrate for coagulation factors IXa and XIIa as well as for trypsin and soybean trypsin-like enzyme.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.