Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Dulbecco's Phosphate Buffered Saline (1X) w/o Calcium and Magnesium

Catalog No. BW17512Q
Change view
Click to view available options
Quantity:
500 mL
1 L
12 x 500 mL
24 x 500 mL
12 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW17-512Q 1 L
BW17512F24 24 x 500 mL
BW17512F12 12 x 500 mL
BW17-512F 500 mL
BW17512Q12 12 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. BW17-512Q Supplier Lonza™ Supplier No. 17512Q

Used to provide buffer system to maintain cell culture media in the physiological pH range of 7 to 7.6.

  • Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
  • Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Dulbecco's Phosphate Buffered Saline
Without Additives No Calcium, No Magnesium
pH 7 to 7.6
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.