Learn More
Cytoskeleton, Inc. Tubulin protein (biotin): porcine brain, 5x20 µg
Supplier:  Cytoskeleton, Inc. T333PA
Encompass
Porcine brain tubulin (>99% pure) has been modified so that random surface lysines contain a covalently linked, long-chain biotin derivative. A long-chain biotin derivative was selected for this procedure because it allows the biotin molecules to be spaced far enough away from the tubulin protein so as not to interfere with its activity, e.g., ligand binding to SPA beads or other streptavidin based reagents. Biotin labeled tubulin is supplied as a lyophilized powder.
Catalog No. NC9123696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.