Learn More
Sigma Aldrich Fine Chemicals Biosciences Biotin, 58-85-5, MFCD00005541, 1g
Supplier: Sigma Aldrich Fine Chemicals Biosciences B45011G
Empirical Formula (Hill Notation) C10H16N2O3S, ≥99% (HPLC), Molecular Weight 244.31, lyophilized powder, Synonym: D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H; Biotin may be used to elute proteins from avidin/streptavidin resins.