Learn More
Biosure Inc BIOSURE 8X SHEATH SOLUTION
Supplier: Biosure Inc 1019
BioSure Sheath Solution is a buffered, azide-free support/carrier reagent for transporting particles through a flow cytometer. The product contains an antibacterial-antifungal preservative.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.