Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 3730/3730xl DNA Analyzer Sequencing Standards, BigDye™ Terminator v3.1

Catalog No. 4336943
Encompass_Preferred
Click to view available options
Quantity:
1 Kit

Catalog No. 43-369-43

Supplier: Applied Biosystems™ 4336943

Only null left
Add to Cart
Edge
Add to Cart

Includes

Kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations

This material is used with the 3730, and 3730xl DNA Analyzers.

This kit contains four tubes of hsp69 standard sequence used for instrument and spectral calibrations.

Order Info

Kit

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains 4 tubes of hsp69 standard sequence used for instrument and spectral calibrations. This material is used with the 3730, and 3730xl DNA Analyzers. Store kit at -15 to -25°C.
Format Tube
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer
For Use With (Application) Sequencing
Product Line BigDye Terminator, BigDye
Product Type Sequencing Standards
Quantity 1 Kit
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.