Learn More
beta L Crystallin Polyclonal Antibody

Description
beta L Crystallin Polyclonal Antibody for ICC/IF, IHC
Specifications
Specifications
| Antigen | beta L Crystallin |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CRYL1 |
| Gene Accession No. | Q9Y2S2 |
| Gene Alias | 1110025H08Rik; A230106J09Rik; C85932; CRY; Cryl1; crystallin lambda 1; crystallin, lambda 1; crystallin, lamda 1; epididymis luminal protein 30; GDH; gul3dh; HEL30; lambda-CRY; Lambda-crystallin homolog; L-gulonate 3-dehydrogenase; testicular tissue protein Li 44 |
| Gene Symbols | CRYL1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.