Learn More
beta-Glycerophosphate, 200mM soln.
Description
β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Specifications
| Color | Colorless |
| Quantity | 50 mL |
| Solubility Information | Soluble in water. |
| Formula Weight | 216.04 |
| Concentration or Composition (by Analyte or Components) | 200mM soln. |
| Physical Form | Liquid |
| Chemical Name or Material | beta-Glycerophosphate |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.