Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Defensin 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981540
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1540 Supplier Bioss Supplier No. BS1163R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.

Specifications

Antigen beta Defensin 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DEFB1
Gene Accession No. P60022
Gene Alias AW260221; BD1; BD-1; beta defensin 1; Beta-defensin 1; beta-defensin-1; DEFB1; DEFB-1; DEFB101; defensin beta 1; Defensin, beta 1; HBD1; rBD-1
Gene Symbols DEFB1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from rat Defensin Beta 1 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1672
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.