Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HemoCue America Beckman Coulter™ Hemoccult™ ICT Immunochemical Fecal Occult Blood Test Kit

Catalog No. 23280011
Click to view available options
Quantity:
100 Pack
40 Pack
Type:
ICT Collection Cards
ICT Sample Collection Kit
This item is not returnable. View return policy

Catalog No. 23280011

Supplier: HemoCue America 395066A

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

ICT FOBT Kit (Cat. No. 23-280-012) includes test devices and 8mL bottle of Hemoccult ICT Buffer; ICT Sample Collection Kit (Cat. No. 23-280-011) contains Patient Kits; each Patient Kit contains Dispensing Envelope with Patient Instructions, Hemoccult ICT Collection Cards, Collection Tissues, Applicator Sticks, and Mailing Pouch; ICT Collection Cards (Cat. No. 23-280-010) contain Collection Cards and Applicator Sticks; ICT Control Kit (Cat. No. 23-280-013) contains bottles (Positive and Negative)

  • CLIA Complexity: Waived
  • CPT Codes: 82274QW, G0328QW
  • Rapid, qualitative immunochemical chromatographic method for detection of human hemoglobin from blood in fecal samples
  • Visually read results

TRUSTED_SUSTAINABILITY

Specifications

CPT Code 82274QW, G0328QW
Clia Complexity Waived
Type ICT Sample Collection Kit
For Use With (Application) Detection of Human hemoglobin from Blood in fecal samples
Includes Dispensing envelope with patient instructions, Hemoccult ICT collection cards, collection tissues, applicator sticks and mailing pouch
Quantity 40 Pack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.