Learn More
BD Cytometric Bead Array Mouse Inflammation Lyophilized Standard
Catalog No. BDB620280 Shop All BD Biosciences Products
Click to view available options
Quantity:
1 Vial
Catalog No. BDB620280
Supplier: BD 620280
Only null left
Add to Cart
Add to Cart
For use as standard control with BD™ Cytometric Bead Array (CBA) Mouse Inflammation Kit
BD™ Cytometric Bead Array Mouse Inflammation Lyophilized Standard contains a mixture of individually lyophilized recombinant proteins made up of recombinant mouse Interleukin-6 (IL-6), IL-10, IL-12p70, Monocyte Chemotractant Protein-1 (MCP-1), Interferon-γ (IFN-γ) and Tumor Necrosis Factor (TNF).
  • Storage buffer: aqueous buffered solution containing BSA and ProClin™ 150
  • Size: 1 vial
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Vial
Product Type Cytometric Bead Array (CBA) Standard
Description Mouse Inflammation Lyophilized Standard
Includes IL-6, IL-10, MCP-1, IFN-gamma, TNF, IL-12p70
Regulatory Status RUO
Content And Storage 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.