Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytofix/Cytoperm™ Fixation and Permeabilization Solution

Catalog No. BDB554722 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
Catalog No. Quantity
BDB554722 125 mL
1 options

Catalog No. BDB554722

Supplier: BD 554722

Only null left
Add to Cart
Add to Cart

1X solution can be used for the simultaneous fixation and permeabilization of cells prior to intracellular cytokine staining

Recommended Assay Procedure:
For the stimulation of cells, various in vitro methods have been reported for stimulating cells to produce cytokines. Polyclonal activators have been particularly useful for inducing cytokine-producing cells. These activators include the following: concanavalin A, lipopolysaccharide, phorbol esters plus calcium ionophore or ionomycin, phytohaemaglutinin, staphylococcus enterotoxin B, and monoclonal antibodies directed against subunits of the TCR/CD3 complex (with or without antibodies directed against costimulatory receptors, such as CD28).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Can be used for the simultaneous fixation and permeabilization of cells prior to intracellular Cyto kine staining
pH Range 7.2
Content And Storage Store undiluted at 4°C
Product Type Fixation and permeabilization solution
Quantity 125 mL
Format Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.