Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ IsoVitaleX™ Enrichment

Catalog No. b11876 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
2 mL
5 x 10 mL
This item is not returnable. View return policy

Catalog No. B11876

Supplier: BD 211876

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

BD BBL™ IsoVitaleX™ Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.

  • GC Agar supplemented with hemoglobin and BBL IsoVitaleX Enrichment (in place of yeast concentrate) provides an improved medium for the isolation of microorganisms
  • Enrichment provides V factor (nicotinamide adenine dinucleotide, NAD) for Haemophilus species
  • Also provides vitamins, amino acids, coenzymes, dextrose, ferric ions and other factors which improve the growth of pathogenic Neisseria
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive IsoVitaleX™ Enrichment
Form Lyophilized
For Use With (Application) Dehydrated culture media
Packaging Type Vial
Quantity 5 x 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.