Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: PPLO Agar (Mycoplasma Agar)

Catalog No. DF0412173 Shop All BD Diagnostic Systems Products
Click to view available options
:
PPLO Agar (Use with Mycoplasma Supplement); 500g
This item is not returnable. View return policy

Catalog No. DF0412-17-3

Supplier: BD 241210

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used with Mycoplasma supplement or Mycoplasma supplement S for isolation and cultivation of Mycoplasma

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type PPLO Agar
Product Type Agar
Formulation Beef heart infusion from 50g 6g/L, Peptone 10g/L, Sodium Chloride 5g/L, Agar 14g/L
For Use With (Application) Used with Mycoplasma supplement or Mycoplasma supplement S for isolation and cultivation of Mycoplasma
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.