Learn More
Gold Biotechnology Inc Bacterial Cell Lysis Buffer
SDP
Supplier:  Gold Biotechnology Inc GB176100G
Encompass
The Bacterial Cell Lysis Kit has been developed for the extraction of biologically active, soluble proteins and inclusion bodies from bacterial cells. The Bacterial Lysis Buffer is a proprietary improvement on the lysozyme based lysis method, which allows extraction of soluble proteins and concurrent removal of nucleic acids (DNA & RNA) released during cell lysis. The kit’s lysis eliminates viscosity build-up, allowing effective clarification with lower centrifugal forces.
Catalog No. NC0964016
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.