Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BACTEC™ PEDS Plus PRIME Medium Culture Vials

Catalog No. b442020 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
50 Vials
This item is not returnable. View return policy

Catalog No. B442020

Supplier: BD 442020

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with BD BACTEC 9000 or BD BACTEC FX Systems. BACTEC™ Bottle Plastic PEDS Plus PRIME Medium Culture Vials requires a software upgrade.

  • Enriched Soybean-Casein Digest broth with CO2
TRUSTED_SUSTAINABILITY

Specifications

Format Plastic Bottle
Product Type Culture Vials
For Use With (Equipment) BD BACTEC 9000 or BD BACTEC FX Systems
Quantity 50 Vials
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.