Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ B-27™ Supplement, minus insulin

Catalog No. A1895601
Encompass_Preferred
Click to view available options
Quantity:
10 mL
100 mL

Catalog No. A1895601

Supplier: Gibco™ A1895601

Only null left
Add to Cart
Edge
Add to Cart

B-27™ Supplement, minus insulin

B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media.

For established protocols using B-27 Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -20°C). Protect from light.
Cell Type Neural Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile-filtered
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility, Mycoplasma
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.