Learn More
Medchemexpress LLC HY-12750 100mg , AZD3965 CAS:1448671-31-5 Purity:98%
Supplier: Medchemexpress LLC HY12750100MG
Medchemexpress, HY-12750 100mg AZD3965 CAS:1448671-31-5 Formula:C21H24F3N5O5S Ki: 1.6 nM (MCT1) Purity:98% AZD3965 is a selective MCT1 inhibitor, it inhibits MCT1 with a binding affinity ( K i ) 1.6 nM and it is 6 fold selective over MCT2 and does not inhibit MCT3 or 4 at 10 μM concentrations. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.