Learn More
Medchemexpress LLC HY-18179 5mg Medchemexpress, AZD2932 CAS:883986-34-3 Purity:>98%
Supplier: Medchemexpress LLC HY181795MG
Medchemexpress, HY-18179 5mg AZD2932 CAS:883986-34-3 AZD2932 is a potent and multi-targeted kinase inhibitor VEGFR2, PDGFβ, Flt-3 and c-Kit with IC50s of 8, 4, 7 and 9 nM in cell assay, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.