Learn More
Medchemexpress LLC HY-19816A 5mg Medchemexpress, Avitinib (maleate) CAS:1557268-88-8 Purity:>98%
Supplier: Medchemexpress LLC HY19816A5MG
Medchemexpress, HY-19816A 5mg Avitinib (maleate) CAS:1557268-88-8 Avitinib maleate is a pyrrolopyrimidine-based irreversible epidermal growth factor receptor (EGFR) inhibitor with an IC50 of 7.68 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.