Learn More
MilliporeSigma™ Supelco™ ATIS Extraction Glassware with Micro Connector
Shop All MilliporeSigma Supelco ProductsDescription
The opening in the glassware will accept solid samples up to 1/2 in. (13 mm) in diameter and up to 3 in. long (76 mm) to be inserted into the glassware. The extraction glassware simply slides into the ATIS heating block. Two types of glass joints are available.
Specifications
Specifications
| For Use With (Application) | Chromatography Applications |