Learn More
Apexbio Technology LLC AT9283, 5mg Cas No: 896466-04-9 MFCD: MFCD13193062
Supplier: Apexbio Technology LLC A41175
AT9283, a synthetic small heterocyclic molecule discovered using a fragment-based approach, is a novel inhibitor of aurora kinase, a family of serine/threonine kinases regulating both mitosis and meiosis, that potently inhibits Aurora kinases A and B. There are other sizes available. Please inqury us for quote.