Learn More
Aqua Communications SnakeEye II™ Video Inspection System
Catalog No. 19806213 Shop All Aqua Communications, Inc Products
Encompass
Click to view available options
Product Type:
Video Inspection System Base Kit
Catalog No. 19-806-213
Supplier: Aqua Communications BK01W
Modular, portable inspection system facilitates quick, nondestructive inspection in hard-to-reach locations
Combines a high-speed CCD camera, built-in LED illumination, and TFT-LCD display technology to deliver high resolution (960 x 234), full color video even in low-light conditions. Choose from two base kits with numerous accessories available.
  • Constructed of heavy-duty, impact- and shock-resistant plastic
  • Designed to resist water, dust, and dirt in harsh industrial and commercial environments
  • Suitable for use where grease and nonsolvent-based chemicals are present
  • Features an interchangeable 1.25 in. diameter camera head that can be attached to a rigid wand, a ring finger attachment or extendable cables
  • Optional C-mount camera adapts to borescopes and fiberscopes
  • Plug-in capabilities allow user to connect to a standard VCR to record and play back images
Specifications
Product Type Video Inspection System Base Kit
Includes LED camera, rigid wand adapter, finger adapter, 4 ft. cable, two NiMH batteries, battery charger, AC adapter, DC auto adapter and a briefcase style carrying case
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.