Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AOF2 Mouse anti-Human, Monkey, Clone:3F9A3, Abnova™

Catalog No. 89334492 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
89-334-492 100 μg
1 options

Catalog No. 89-334-492

Supplier: Abnova Corporation MAB17849

Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human AOF2.

This gene encodes a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Specifications

Antigen AOF2
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3F9A3
Conjugate Unconjugated
Description amine oxidase (flavin containing) domain 2
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Immunofluorescence (1:200-1000)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene AOF2
Gene Alias BHC110, KDM1, KIAA0601, LSD1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 55-263 of human AOF2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 23028
Target Species Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.