Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSLPR/CRLF2 Antibody, Novus Biologicals™
SDP

Mouse Polyclonal Antibody

$149.33 - $480.74

Specifications

Antigen TSLPR/CRLF2
Dilution Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5-7 ug/ml
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP222383SS
SDP
View Documents
Novus Biologicals
NBP222383SS
0.025 mg
Each for $149.33
Only null left
Add to Cart
 
NBP222383
SDP
View Documents
Novus Biologicals
NBP222383
0.1 mg
Each for $480.74
Only null left
Add to Cart
 
Description

Description

TSLPR/CRLF2 Polyclonal specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TSLPR/CRLF2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
AF338733_1
64109
Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5-7 ug/ml
Polyclonal
Purified
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR
CRLF2
IgG2a κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.