Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sodium channel, voltage-gated, type IX, alpha subunit, Mouse, Clone: 5A11, Abnova™

Catalog No. 89014186 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
89-014-186 100 μg
1 options

Catalog No. 89-014-186

Supplier: Abnova Corporation H00006335M01

Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SCN9A.

This gene encodes a voltage-gated sodium channel which plays a significant role in nociception signaling. Mutations in this gene have been associated with primary erythermalgia, channelopathy-associated insensitivity to pain, and paroxysmal extreme pain disorder. [provided by RefSeq

Sequence: GNLKHKCFRNSLENNETLESIMNTLESEEDFRKYFYYLEGSKDALLCGFSTDSGQCPEGYTCVKIGRNPDY

Specifications

Antigen sodium channel, voltage-gated, type IX, alpha subunit
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SCN9A.
Formulation PBS with no preservative; pH 7.4
Gene SCN9A
Gene Accession No. NM_002977
Gene Alias ETHA/NE-NA/NENA/Nav1.7/PN1
Gene Symbols SCN9A
Host Species Mouse
Immunogen SCN9A (NP_002968, 269 a.a. ∼ 339 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6335
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.