Learn More
receptor-interacting serine-threonine kinase 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RIPK2 protein.
Supplier: Abnova Corporation H00008767B01
Description
This gene encodes a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases. The encoded protein contains a C-terminal caspase activation and recruitment domain (CARD), and is a component of signaling complexes in both the innate and adaptive immune pathways. It is a potent activator of NF-kappaB and inducer of apoptosis in response to various stimuli. [provided by RefSeq
Sequence: MNGEAICSALPTIPYHKLADLRYLSRGASGTVSSARHADWRVQVAVKHLHIHTPLLDSERKDVLREAEILHKARFSYILPILGICNEPEFLGIVTEYMPNGSLNELLHRKTEYPDVAWPLRFRILHEIALGVNYLHNMTPPLLHHDLKTQNILLDNEFHVKIADFGLSKWRMMSLSQSRSSKSAPEGGTIIYMPPENYEPGQKSRASIKHDIYSYAVITWEVLSRKQPFEDVTNPLQIMYSVSQGHRPVINEESLPYDIPHRARMISLIESGWAQNPDERPSFLKCLIELEPVLRTFEEITFLEAVIQLKKTKLQSVSSAIHLCDKKKMELSLNIPVNHGPQEESCGSSQLHENSGSPETSRSLPAPQDNDFLSRKAQDCYFMKLHHCPGNHSWDSTISGSQRAAFCDHKTTPCSSAIINPLSTAGNSERLQPGIAQQWIQSKREDIVNQMTEACLNQSLDALLSRDLIMKEDYELVSTKPTRTSKVRQLLDTTDIQGEEFAKVIVQKLKDNKQMGLQPYPEILVVSRSPSLNLLQNKSMSpecifications
| receptor-interacting serine-threonine kinase 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RIPK2 protein. | |
| RIPK2 | |
| CARD3/CARDIAK/CCK/GIG30/RICK/RIP2 | |
| Mouse | |
| 50 μL | |
| Apoptosis | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_003821.5 | |
| RIPK2 | |
| RIPK2 (NP_003812.1, 1 a.a. ∼ 540 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 8767 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.