Learn More
Invitrogen™ PTGIR Polyclonal Antibody

Description
PA1-32775 detects Prostacyclin receptor from human samples.
Specifications
Specifications
| Antigen | PTGIR |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | PTGIR |
| Gene Accession No. | P43119 |
| Gene Alias | IP; IP receptor; PGI receptor; PGI2; PGI2 receptor; PRIPR; Prostacyclin receptor; prostaglandin I receptor (IP); prostaglandin I2 (prostacyclin) receptor (IP); prostaglandin I2 receptor; Prostanoid IP receptor; Ptgir |
| Gene Symbols | PTGIR |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.