Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K(lysine) acetyltransferase 2B, Mouse, Clone: 1H2, Abnova™

Catalog No. 89014228 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
89-014-228 100 μg
1 options

Catalog No. 89-014-228

Supplier: Abnova Corporation H00008850M02

Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PCAF.

CBP and p300 are large nuclear proteins that bind to many sequence-specific factors involved in cell growth and/or differentiation, including c-jun and the adenoviral oncoprotein E1A. The protein encoded by this gene associates with p300/CBP. It has in vitro and in vivo binding activity with CBP and p300, and competes with E1A for binding sites in p300/CBP. It has histone acetyl transferase activity with core histones and nucleosome core particles, indicating that this protein plays a direct role in transcriptional regulation. [provided by RefSeq

Sequence: LEEEVYSQNSPIWDQDFLSASSRTSQLGIQTVINPPPVAGTISYNSTSSSLEQPNAGSSSPACKASSGLEANPGEKRKMTDSHVLEEAKKPRVMGDIPME

Specifications

Antigen K(lysine) acetyltransferase 2B
Applications ELISA, KnockDown, Western Blot
Classification Monoclonal
Clone 1H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PCAF.
Formulation PBS with no preservative; pH 7.4
Gene KAT2B
Gene Accession No. BC060823
Gene Alias CAF/P/P/CAF/PCAF
Gene Symbols KAT2B
Host Species Mouse
Immunogen PCAF (AAH60823, 353 a.a. ∼ 452 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 8850
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.