Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KNP-1 Mouse, Unlabeled, Clone: 35, BD

Catalog No. p-3850303 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611597 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611597 Supplier BD Biosciences Supplier No. 611597

Mouse Monoclonal Antibody

Human chromosome 21 has been extensively studied because trisomy 21 causes Down syndrome. In addition to this syndrome, chromosome 21 is the loci for five other hereditary disorders, myoclonus epilepsy, autoimmune polyglandular disease type I, nonsyndromic hereditary deafness, Knobloch syndrome, and bipolar affective disorder. Exon trapping studies of human chromosome 21q22.3 identified transcriptional units with homology to the zebrafish ES1 and the E. coli sigma cross-reacting protein 27A (SCRP27A). The human ES1 homolog (HES1) was also identified as KNP-1α /KNP-1β, as well as GT335 in similar screens for genes involved in disorders that map to chromosome 21q22.3. KNP-1/HES1 mRNA is ubiquitously expressed with the highest expression in muscle and heart. KNP-1/HES1 protein contains putative mitochondrial targeting signals and localizes to mitochondria in mouse NIH 3T3 cells. Interestingly, mitochondrial deletions have been implicated in syndromes related to deafness. Thus, KNP-1/HES1 may be a mitochondrial protein important for normal development, and may be a candidate gene for disorders related to deletions at chromosome 21q22.3.

Host Species: Mouse
Clone: 35
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human ES1 aa. 49-240

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen KNP-1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 35
Concentration 250μg/mL
Conjugate Unconjugated
Description HES1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ES1 aa. 49-240
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.