Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
glutamine-fructose-6-phosphate transaminase 2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human GFPT2 protein.
Supplier: Abnova Corporation H00009945B01P
Description
Sequence: MCGIFAYMNYRVPRTRKEIFETLIKGLQRLEYRGYDSAGVAIDGNNHEVKERHIQLVKKRGKVKALDEELYKQDSMDLKVEFETHFGIAHTRWATHGVPSAVNSHPQRSDKGNEFVVIHNGIITNYKDLRKFLESKGYEFESETDTETIAKLIKYVFDNRETEDITFSTLVERVIQQLEGAFALVFKSVHYPGEAVATRRGSPLLIGVRSKYKLSTEQIPILYRTCTLENVKNICKTRMKRLDSSACLHAVGDKAVEFFFASDASAIIEHTNRVIFLEDDDIAAVADGKLSIHRVKRSASDDPSRAIQTLQMELQQIMKGNFSAFMQKEIFEQPESVFNTMRGRVNFETNTVLLGGLKDHLKEIRRCRRLIVIGCGTSYHAAVATRQVLEELTELPVMVELASDFLDRNTPVFRDDVCFFISQSGETADTLLALRYCKDRGALTVGVTNTVGSSISRETDCGVHINAGPEVGVASTKAYTSQFISLVMFGLMMSEDRISLQNRRQEIIRGLRSLPELIKEVLSLEEKIHDLALELYTQRSLLVMGRGYNYATCLEGALKIKEITYMHSEGILAGELKHGPLALIDKQMPVIMVIMKDPCFAKCQNALQQVTARQGRPIILCSKDDTESSKFAYKTIELPHTVDCLQGILSVIPLQLLSFHLAVLRGYDVDFPRNLAKSVTVESpecifications
| glutamine-fructose-6-phosphate transaminase 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human GFPT2 protein. | |
| GFPT2 | |
| FLJ10380/GFAT2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000012.2 | |
| GFPT2 | |
| GFPT2 (AAH00012.1, 1 a.a. ∼ 682 a.a) full-length human protein. | |
| 50 μg | |
| Stem Cell Biology | |
| 9945 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction