Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR7/RDC-1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$206.50 - $519.67

Specifications

Antigen CXCR7/RDC-1
Concentration 1.0 mg/mL
Dilution Western Blot 4 - 6 ug/mL, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Flow Cytometry (Negative)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224779SS
SDP
View Documents
Novus Biologicals
NBP224779SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224779
SDP
View Documents
Novus Biologicals
NBP224779
0.1 mg
Each for $519.67
Only null left
Add to Cart
 
Description

Description

CXCR7/RDC-1 Polyclonal specifically detects CXCR7/RDC-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow Cytometry (Negative).
Specifications

Specifications

CXCR7/RDC-1
Western Blot 4 - 6 ug/mL, Simple Western 1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 - 10 ug/mL, Flow Cytometry (Negative)
Polyclonal
Purified
RUO
Human, Mouse, Rat, Sheep
P25106
57007
A synthetic peptide corresponding amino acids 106-129 of human CXCR7/RDC1 was used as the immunogen, GenBank NP_064707.1.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
Unconjugated
Rabbit
Chemokines and Cytokines, GPCR
PBS with 0.05% Sodium Azide
chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
ACKR3
IgG
Peptide affinity purified
This antibody is reactive to Chimpanzee, Cow, Horse, Human, Monkey, Mouse, Rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.