Learn More
MilliporeSigma™ anti-Brain Lipid-Binding Protein (BLBP), Polyclonal, ™
Shop All MilliporeSigma Products
Description
BLBP (Brain lipid-binding protein) belongs to a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. BLBPs play essential roles in fatty acid uptake, transport and metabolism. BLBP can be used as a molecular marker for radial glia (a major neural progenitor cell type and a scaffolding supporting neuronal migration) in the developing central nervous system. Radial glia play an essential role in the generation of the cerebral cortex through their function as neuronal precursors and as neuronal migration guides.
Host Species: Rabbit
Cross Reactivity: Human, Mouse
Immunogen: GST-tagged recombinant protein corresponding to human Brain lipid-binding protein.
Immunohistochemistry, Western Blotting
Specifications
Specifications
| Antigen | Brain Lipid-Binding Protein (BLBP) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | GST-tagged recombinant protein corresponding to human Brain lipid-binding protein. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Target Species | Human, Mouse |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.