Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Andwin Scientific 100-Slide Capacity Microscope Slide Storage Boxes
Click to view available options
Color:
Blue
Red
White
Yellow
Description
- Constructed from polystyrene
- 100-Slide capacity
- Corrosion-resistant hinges and clasp
- Cork-lined bottoms
- Individually numbered slide positions
- Available in four colors
Specifications
Specifications
| Slide Capacity | 100 |
| Color | Yellow |
| Device Holds | 100 |
| For Use With (Equipment) | Microscope Slide |
| Includes | Cork lined bottom |
| Material | Polystyrene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction