Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andwin Scientific 100-Slide Capacity Microscope Slide Storage Boxes

Catalog No. 1437285
Click to view available options
Color:
Blue
Red
White
Yellow

Catalog No. 14-372-85

Supplier: Andwin Scientific 880100

Only null left
Add to Cart
Add to Cart

Designed for safe, long-term slide storage

  • Constructed from polystyrene
  • 100-Slide capacity
  • Corrosion-resistant hinges and clasp
  • Cork-lined bottoms
  • Individually numbered slide positions
  • Available in four colors

Specifications

Slide Capacity 100
Color Red
Device Holds 100
For Use With (Equipment) Microscope Slide
Includes Cork lined bottom
Material Polystyrene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.