Learn More
Enzo Life Sciences Anacardic acid (25mg). CAS: 16611-84-0
Supplier: Enzo Life Sciences ALX270381M025
Cell permeable salicylic acid analog that acts as a potent, non-competitive inhibitor of p300 and PCAF (p300/CBP-associated factor) histone acetyltransferase (HAT) activities (IC50~8.5µM and ~5µM, respectively). Displays a variety of other biological activities, such as antibacterial, antimicrobial, prostaglandin synthase inhibition, tyrosinase, and lipoxygenase inhibition.Inhibits SUMOylation of RanGAP1-C2 with an IC50 = 2.2µM Alternative name: 2-Hydroxy-6-pentadecylbenzoic acid, 6-Pentadecylsalicylic acid, AA. Formula: C22H36O3. MW: 348.5. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol, DMSO, dichloromethane or ethylacetate. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.