Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMOTL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578770
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA578770 100 μg
1 options

Catalog No. PIPA578770

Supplier: Invitrogen™ PA578770

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, rat brain tissue, rat lung tissue, mouse lung tissue, mouse liver tissue.

Regulates the translocation of phosphorylated SRC to peripheral cell-matrix adhesion sites. Required for proper architecture of actin filaments. Inhibits the Wnt/beta-catenin signaling pathway, probably by recruiting CTNNB1 to recycling endosomes and hence preventing its translocation to the nucleus. Participates in angiogenesis. May play a role in the polarity, proliferation and migration of endothelial cells. Selectively promotes FGF-induced MAPK activation through SRC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMOTL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.01mg sodium azide
Gene AMOTL2
Gene Accession No. G3V735, Q8K371, Q9Y2J4
Gene Alias AMOTL2; angiomotin like 2; angiomotin-like 2; angiomotin-like protein 2; AW549739; C79691; KIAA0989; LCCP; Leman coiled-coil protein; MASCOT; mKIAA0989
Gene Symbols AMOTL2
Host Species Rabbit
Immunogen E. coli-derived human AMOTL2 recombinant protein (Position: A401-Q480).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51421, 56332, 65157
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.