Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Sulfide Solution, Reagent, ACS, 20-24%, Spectrum™ Chemical
SDP

Catalog No. 18600670 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
6 x 500 mL
20 L
Packaging:
Amber Glass Bottle
Poly-Coated Glass Bottle
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-670 500 mL Amber Glass Bottle
18-600-669 100 mL Amber Glass Bottle
18-600-672 2.5 L Poly-Coated Glass Bottle
18-600-671 6 x 500 mL Amber Glass Bottle
18-600-673 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-670 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1404500MLGL
Only null left
Add to Cart
Add to Cart

A1404, 12135-76-1, (NH4)2S

Spectrum™ Chemical Ammonium Sulfide Solution, Reagent, ACS is an unstable salt with the formula (NH4)2S. Common uses of AMMONIUM SULFIDE are industrial applications including photo developing and textile manufacturing. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 12135-76-1 , 7732-18-5
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

CAS Max % 80%, 20%
Quantity 500 mL
UN Number UN2683
Percent Purity 20 to 24%
Grade ACS
Packaging Amber Glass Bottle
Residue after Ignition 0.04%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.