Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMG 333
GREENER_CHOICE

Catalog No. 687410
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 68-741-0

Supplier: Tocris Bioscience™ 6874/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective TRPM8 antagonist

AMG 333 is a potent and selective TRPM8 antagonist (IC50 values are 13 and 20 nM for hTRPM8 and rTRPM8, respectively). Displays high selectivity over other TRP channels (IC50 >20 μM, TRPV1/V3/V4/A1). Inhibits icilin -induced wet-dog shake in rats and suppresses cold-induced increases in blood pressure in the rat cold-pressor test.

Specifications

Quantity 10 mg
Molecular Formula C20H12F5N3O4
Formula Weight 453.33
Product Type Antagonist
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.