Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TE, pH 7.0, RNase-free

Catalog No. am9861
Encompass_Preferred
Click to view available options
Quantity:
50 mL
10 x 1 mL

Catalog No. AM9861

Supplier: Invitrogen™ AM9861

Only null left
Add to Cart
Add to Cart

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water.

Invitrogen TE buffer is a commonly recommended storage solution for RNA samples. 10 mM Tris-HCl, 1 mM EDTA, pH 7.0 is in nuclease-free, ultrapure water. The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. TE Buffer is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

pH 7.0
Physical Form Liquid
Product Line Ambion
Quantity 50 mL
Packaging Bottle

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.