Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 10X TBE (powder)

Catalog No. am9864
Encompass_Preferred
Click to view available options
Quantity:
10 x 1 L

Catalog No. AM9864

Supplier: Invitrogen™ AM9864

Only null left
Add to Cart
Edge
Add to Cart

Invitrogen 10X TBE (Tris-Borate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitive application.

Invitrogen 10X TBE (Tris-Borate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitive application. 10 packets are provided, each containing the necessary powder to make 1 L of 10X TBE buffer upon the addition of H2O. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Running Buffer
Concentration 10X
For Use With (Application) Nucleic acid gel electrophoresis & blotting
Physical Form Powder
Product Line Ambion
Quantity 10 x 1 L
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.