Learn More
Description
- Suitable for Level 4 Biological Safety Laboratories
- Provides protection against chemicals in liquid, aerosol and solid form
- Protects against infectious agents, i.e. bacteria, virus and fungi, and against radioactive particles
- Designed for use with an external air source, which allows longer working hours and provides improved wearer comfort
- Made from a strong yet soft and durable material
Specifications
Specifications
| Color | Orange |
| For Use With (Application) | Biosafety Labs Levels 1-4, Centers for Disease Control and Prevention, Life Sciences, Chemical, Mining, Fire and Rescue, Oil and Gas |
| Material | Polyester |
| Size | Small (Suit), 6 (Socks) |
| Closure Type | Heavy-duty vapor-protective zipper with D-rings |
| Disposable | Reusable |
| Product Type | Chemical Protective Suits |
| Seam Type | Stitched and welded |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.